Learn More
Medchemexpress LLC 2-Oxazolidinone, 3-(1-oxo-2-buten-1-yl)-4-(phenylmethyl)-, (4S)- | 133812-16-5 | 99.5% | 245.27 | 100 MG
SDP
Catalog No. 5000454568
Encompass_Preferred
(E/Z)-Locostatin ((E/Z)-UIC-1005) is a racemic form of Locostatin. Locostatin (UIC-1005) acts as a potent RKIP inhibitor. It functions by binding to the Raf kinase inhibitor RKIP protein, thereby disrupting the interaction between RKIP and Raf-1 kinase, as well as G protein-coupled receptor kinase 2. Locostatin has been shown to inhibit cell proliferation and migration. Additionally, it has been observed to aggravate thioacetamide (HY-Y0698)-induced acute liver failure in mice.
  • Racemic form of locostatin
  • Potent RKIP inhibitor
  • Disrupts interaction of RKIP with raf-1 kinase and G protein-coupled receptor kinase 2
  • Inhibits cell proliferation and migration
Catalog No. 50-004-54568 Supplier Medchemexpress LLC Supplier No. HYW013411100MG
(E/Z)-Locostatin ((E/Z)-UIC-1005) is a racemic form of Locostatin. Locostatin (UIC-1005) acts as a potent RKIP inhibitor. It functions by binding to the Raf kinase inhibitor RKIP protein, thereby disrupting the interaction between RKIP and Raf-1 kinase, as well as G protein-coupled receptor kinase 2. Locostatin has been shown to inhibit cell proliferation and migration. Additionally, it has been observed to aggravate thioacetamide (HY-Y0698)-induced acute liver failure in mice.
  • Racemic form of locostatin
  • Potent RKIP inhibitor
  • Disrupts interaction of RKIP with raf-1 kinase and G protein-coupled receptor kinase 2
  • Inhibits cell proliferation and migration