Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Oxazolidinone, 4-(1-methylethyl)-, (4R)- | 95530-58-8 | 129.16 | 25 G
SDP

Catalog No. 5000448110
Encompass_Preferred

(R)-4-Isopropyloxazolidin-2-one is a chemical compound offered by MedChemExpress, identified by catalog number HY-W002320. This product is rigorously tested to ensure compliance with its specifications, making it suitable for various laboratory applications requiring high-purity reagents.

  • White to off-white solid appearance
  • Purity of 99.78% via GC analysis
  • Structure confirmed by ¹H NMR spectroscopy
  • Recommended storage: -20°C for powder up to 3 years; -80°C in solvent up to 6 months

Catalog No. 50-004-48110 Supplier Medchemexpress LLC Supplier No. 25GHYW002320
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(R)-4-Isopropyloxazolidin-2-one is a chemical compound offered by MedChemExpress, identified by catalog number HY-W002320. This product is rigorously tested to ensure compliance with its specifications, making it suitable for various laboratory applications requiring high-purity reagents.

  • White to off-white solid appearance
  • Purity of 99.78% via GC analysis
  • Structure confirmed by ¹H NMR spectroscopy
  • Recommended storage: -20°C for powder up to 3 years; -80°C in solvent up to 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.