Learn More
Medchemexpress LLC 2-Oxazolidinone-4,4,5-d3, 3-amino-5-(4-morpholinylmethyl-d2)- | 1017793-94-0 | 99.0% | 25 MG
SDP
Catalog No. 5000339518
Encompass_Preferred
AMOZ-d5 is a deuterium labeled AMOZ. AMOZ is described as a tissue-bound metabolite of Furaltadone, which is a widely used synthetic nitrofuran antibiotic. This product is intended for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-39518 Supplier Medchemexpress LLC Supplier No. 25MGHY131144S
AMOZ-d5 is a deuterium labeled AMOZ. AMOZ is described as a tissue-bound metabolite of Furaltadone, which is a widely used synthetic nitrofuran antibiotic. This product is intended for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS