Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Oxo-2,3-dihydro-1,3-benzothiazole-6-sulfonyl chloride

Catalog No. CDS002659 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002659 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002659 Supplier Sigma Aldrich Supplier No. CDS00265950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,2,2α-trimethylpropionanilide (2,2,2-n-pivaloyl-o-toluidine) may be used to determine the concentration of lithium diisopropylamide mono(tetrahydrofuran) via titration
  • It may be used to determine the concentrations of alkyllithiums by titration, to the colorimetric endpoint, indicated by the persistence of a yellow color in the solution

Specifications

Molecular Formula C7H4ClNO3S2
Linear Formula C7H4ClNO3S2
MDL Number MFCD03033469
Quantity 5 g
Molecular Weight (g/mol) 249.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.