Learn More
Medchemexpress LLC 2-oxo-2H-1-benzopyran-3-carboxylic acid | 531-81-7 | MFCD00006852 | 99.9% | 190.15 g/mol | C10H6O4 | 50 G
SDP
Catalog No. 5000301711
Encompass_Preferred
Coumarin-3-carboxylic acid is a small-molecule research reagent (2-oxochromene-3-carboxylic acid) used as a fluorescent probe for hydroxyl radicals and as a synthetic building block. Supplied as a high-purity solid, it is suitable for in vitro studies and can be formulated for in vivo dosing when required.
Catalog No. 50-003-01711 Supplier Medchemexpress LLC Supplier No. HYW02796850G
Coumarin-3-carboxylic acid is a small-molecule research reagent (2-oxochromene-3-carboxylic acid) used as a fluorescent probe for hydroxyl radicals and as a synthetic building block. Supplied as a high-purity solid, it is suitable for in vitro studies and can be formulated for in vivo dosing when required.