Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxo-4-phenylbutyric Acid 98.0+%, TCI America™
SDP

Catalog No. O03895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03895G Supplier TCI America Supplier No. O03895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 710-11-2
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00969471
InChI Key PPKAIMDMNWBOKN-UHFFFAOYSA-N
PubChem CID 69732
IUPAC Name 2-oxo-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCC(=O)C(=O)O

Specifications

Melting Point 49°C
Color Yellow
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Oxo-4-phenylbutyric Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.