Learn More
Cayman Chemical 2-Oxoadipic Acid, Purity: ≥95%, CAS: 3184-35-8, 50 mg
SDP
Catalog No. 502949228
Encompass_Preferred
An intermediate in the catabolism of L-tryptophan, L-lysine, and hydroxylysine; urinary excretion is increased in patients with α-ketoadipic aciduria
Catalog No. 50-294-9228 Supplier Cayman Chemical Supplier No. 3457450 MG
An intermediate in the catabolism of L-tryptophan, L-lysine, and hydroxylysine; urinary excretion is increased in patients with α-ketoadipic aciduria