Learn More
Medchemexpress LLC HY-W007926 1g Medchemexpress, 2-Oxobutanoic acid CAS:600-18-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0079261G
Encompass_Preferred
Medchemexpress, HY-W007926 1g 2-Oxobutanoic acid CAS:600-18-0 2-Oxobutanoic acid is a product in the enzymatic cleavage of cystathionine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4502