Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Oxovaleric acid | 1821-02-9 | 95.0% | 116.12 | 50 MG
SDP

Catalog No. 5000437995
Encompass_Preferred

2-Oxovaleric acid is a keto acid found in human blood. This product is intended for research and analytical applications, and is for research use only.

  • Analytical standard for 2-Oxovaleric acid
  • Suitable for research and analytical applications
  • Endogenous metabolite
  • Purity of 95.0%
  • Molecular weight of 116.12
  • Supplied as a liquid

Catalog No. 50-004-37995 Supplier Medchemexpress LLC Supplier No. HY11309850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Oxovaleric acid is a keto acid found in human blood. This product is intended for research and analytical applications, and is for research use only.

  • Analytical standard for 2-Oxovaleric acid
  • Suitable for research and analytical applications
  • Endogenous metabolite
  • Purity of 95.0%
  • Molecular weight of 116.12
  • Supplied as a liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.