Learn More
O Berk Company 2OZ FLINT WIDE MOUTH SQUARE GLASS BOTTLE - 33-400 NECK 144/pk
Catalog No. NC9538613
Encompass_Preferred
Glass Bottle: 2OZ Flint Glass Wide Mouth Square with 33-400 Neck Finish; Size 2.00 oz; Overflow Capacity 87.24 ml; Neck Diameter 33.00; Neck Finish 400; Material Glass; Color Flint; Shape Square; Item Weight 77.96 g; Item Height 3.30 in; Item Length 2.00 in; Item Width / Diameter 1.60 in 144/pk
Catalog No. NC9538613 Supplier O Berk Company Supplier No. 23220017
Glass Bottle: 2OZ Flint Glass Wide Mouth Square with 33-400 Neck Finish; Size 2.00 oz; Overflow Capacity 87.24 ml; Neck Diameter 33.00; Neck Finish 400; Material Glass; Color Flint; Shape Square; Item Weight 77.96 g; Item Height 3.30 in; Item Length 2.00 in; Item Width / Diameter 1.60 in 144/pk