Learn More
Bellco Glass, Inc. 2-P Stainless Steel Sparger Assembly , 36L STD Over Head Drive Vessels - Air Stone
SDP
Catalog No. 501944798
Encompass_Preferred
2-port air stone stainless steel sparger assembly is designed for the sparging of gases into the vessel liquid media. This design allows access through the side arms with a screw on cap assembly. Fully autoclavable. MADE IN THE USA.
Catalog No. 50-194-4798 Supplier Bellco Glass, Inc. Supplier No. 196445036R
2-port air stone stainless steel sparger assembly is designed for the sparging of gases into the vessel liquid media. This design allows access through the side arms with a screw on cap assembly. Fully autoclavable. MADE IN THE USA.