Learn More
Bellco Glass, Inc. 2-P Stainless Steel Sparger Assembly , 8-15L STD Over Head Drive Vessels - Air Stone
SDP
Catalog No. 501944797
Encompass_Preferred
2-Port Air Stone Stainless Steel Sparger Assembly is designed for the sparging of gases into the vessel liquid media. This design allows access through the side arms with a screw on cap assembly. Fully autoclavable. MADE IN THE USA. Specifications:Sparger Assembly For 6L L/P OHD VesselFits Thru 45mm Sidearm316L SST Sparger Tube Is 15″ Long With 12mm Glass Frit. Course Porosity 40-60 Microns32 Ra Material Finish Or Better
Catalog No. 50-194-4797 Supplier Bellco Glass, Inc. Supplier No. 196445008R
2-Port Air Stone Stainless Steel Sparger Assembly is designed for the sparging of gases into the vessel liquid media. This design allows access through the side arms with a screw on cap assembly. Fully autoclavable. MADE IN THE USA. Specifications:Sparger Assembly For 6L L/P OHD VesselFits Thru 45mm Sidearm316L SST Sparger Tube Is 15″ Long With 12mm Glass Frit. Course Porosity 40-60 Microns32 Ra Material Finish Or Better