Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(P-Sulfophenylazo)-1,8-dihydroxy-3,6-naphthalenesulfonic acid trisodium salt, MP Biomedicals™

Catalog No. ICN15672910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15672910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15672910 Supplier MP Biomedicals, Inc Supplier No. 0215672910
Add to Cart
Add to Cart

Reagent used for analysis of fluoride, zirconium and thorium

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23647-14-5
Molecular Formula C16H10N2Na3O11S3+
Molecular Weight (g/mol) 571.408
MDL Number MFCD00003952
InChI Key LSAWBVPBZOEGOT-UHFFFAOYSA-L
Synonym spadns, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-4-sulfophenyl azo-, trisodium salt, trisodium 4,5-dihydroxy-3-e-2-4-sulfonatophenyl diazen-1-yl naphthalene-2,7-disulfonate, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-2-4-sulfophenyl diazenyl-, sodium salt 1:3, 2-p-sulfophenylazo-1,8-dihydroxy-3,6-naph-thalenedisulfonic acid
PubChem CID 73658865
IUPAC Name trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]

Specifications

Color Maroon
Quantity 10 g
Formula Weight 570.41
Physical Form Powder
Chemical Name or Material 2-(P-Sulfophenylazo)-1, 8-dihydroxy-3, 6-naphthalenesulfonic acid trisodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.