Learn More
Azzota Corp 2"" (51mm) Diameter Hollow Cathode Lamp For Perkin Elmer Atomic Absorption. Lead - Pb
Catalog No. 501612990
Encompass_Preferred
2" (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption. Lead - Pb
Catalog No. 50-161-2990 Supplier Azzota Corp Supplier No. SHC20PB
2" (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption. Lead - Pb