Learn More
MSC PEC Tools Combination Square Set, 300mm (12 in.), Cast Iron, 1 ea.
Catalog No. 5000513551
Encompass
This 2-piece combination square set is a versatile, professional-grade tool designed for precise layout, measurement, and 90°/45° calibration in machining, woodworking, and metal fabrication. The set includes a precision-machined square/miter head and a durable blade, both engineered for high-accuracy applications.
  • Includes a fully hardened and tempered machinist's blade with a glare-reducing, rust-resistant satin chrome finish.
  • Features a square/miter head equipped with an integrated level vial and a hardened scriber for marking workpieces.
  • Utilizes high-contrast, micro-fine blade graduations in E/M (32nds, 64ths, .5mm, and 1mm) for clear and accurate readings.
  • Constructed with a durable, enamelled blue or hammertone powder-coat finish on non-ground surfaces for long-term wear resistance.
  • Supplied in a fitted blow-moulded case for organized storage and protection.
Catalog No. 50-005-13551 Supplier MSC Supplier No. 06504112
This 2-piece combination square set is a versatile, professional-grade tool designed for precise layout, measurement, and 90°/45° calibration in machining, woodworking, and metal fabrication. The set includes a precision-machined square/miter head and a durable blade, both engineered for high-accuracy applications.
  • Includes a fully hardened and tempered machinist's blade with a glare-reducing, rust-resistant satin chrome finish.
  • Features a square/miter head equipped with an integrated level vial and a hardened scriber for marking workpieces.
  • Utilizes high-contrast, micro-fine blade graduations in E/M (32nds, 64ths, .5mm, and 1mm) for clear and accurate readings.
  • Constructed with a durable, enamelled blue or hammertone powder-coat finish on non-ground surfaces for long-term wear resistance.
  • Supplied in a fitted blow-moulded case for organized storage and protection.