Learn More
MSC PEC Tools Combination Square Set, 6 in., Hardened Steel, 2 Pieces, 1 ea
Catalog No. 5000513308
Encompass
This 2-piece combination square set is engineered for high-precision measurement and layout applications. The set includes a 6 in. hardened steel blade with high-contrast, micro-fine graduations and a square/miter head equipped with a level vial and a hardened scribe tool.
  • Constructed from fully hardened and tempered steel for durability
  • Features a satin chrome finish on blades for improved visibility
  • Includes a square/miter head with integrated level vial and scribe tool
  • Provides high-contrast, micro-fine graduations in 0.5 mm, 1 mm, 32nds, and 64ths
  • Heads supplied with a durable hammertone powder-coat finish
Catalog No. 50-005-13308 Supplier MSC Supplier No. 06504054
This 2-piece combination square set is engineered for high-precision measurement and layout applications. The set includes a 6 in. hardened steel blade with high-contrast, micro-fine graduations and a square/miter head equipped with a level vial and a hardened scribe tool.
  • Constructed from fully hardened and tempered steel for durability
  • Features a satin chrome finish on blades for improved visibility
  • Includes a square/miter head with integrated level vial and scribe tool
  • Provides high-contrast, micro-fine graduations in 0.5 mm, 1 mm, 32nds, and 64ths
  • Heads supplied with a durable hammertone powder-coat finish