Learn More
MSC PEC Tools Combination Square Set, 18 in., 2 Piece, 1 ea.
Catalog No. 5000513228
Encompass
This 2-piece combination square set includes an 18 in. hardened and tempered machinist's blade with a satin chrome finish and a square/miter head. The set is designed for versatile layout and measurement tasks in metalworking and machining environments.
  • Features 4R graduations of 8ths, 16ths, 32nds and 64ths
  • Square head and blade are fully hardened and tempered for durability
  • Square/miter head includes an integrated level vial and hardened scribe tool
  • Heads are finished with a durable hammertone powder-coat
  • High-contrast, micro-fine blade graduations provide easy reading
  • Manufactured in the U.S.A.
Catalog No. 50-005-13228 Supplier MSC Supplier No. 06504138
This 2-piece combination square set includes an 18 in. hardened and tempered machinist's blade with a satin chrome finish and a square/miter head. The set is designed for versatile layout and measurement tasks in metalworking and machining environments.
  • Features 4R graduations of 8ths, 16ths, 32nds and 64ths
  • Square head and blade are fully hardened and tempered for durability
  • Square/miter head includes an integrated level vial and hardened scribe tool
  • Heads are finished with a durable hammertone powder-coat
  • High-contrast, micro-fine blade graduations provide easy reading
  • Manufactured in the U.S.A.