Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentadecanone, 97%

Catalog No. AAL0934722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0934722 100 g
AAL0934714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0934722 Supplier Thermo Scientific Chemicals Supplier No. L0934722
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2345-28-0 | C15H30O | 226.404 g/mol

It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.

Solubility
Soluble in toluene (almost transparency). Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2345-28-0
Molecular Formula C15H30O
Molecular Weight (g/mol) 226.404
MDL Number MFCD00053712
InChI Key CJPNOLIZCWDHJK-UHFFFAOYSA-N
Synonym 2-pentadecanone, methyl tridecyl ketone, pentadecanone, unii-b2q48j997n, fema no. 3724, 2-pentandecanone, acmc-209g3f, 2-pentadecanone, fg, 2-pentadecanone, analytical standard
PubChem CID 61303
IUPAC Name pentadecan-2-one
SMILES CCCCCCCCCCCCCC(=O)C

Specifications

Melting Point 37°C to 41°C
Density 0.82
Boiling Point 293°C to 294°C
Flash Point 144°C (291°F)
Quantity 100 g
Beilstein 1764872
Solubility Information Soluble in toluene (almost transparency). Insoluble in water.
Formula Weight 226.4
Percent Purity 97%
Chemical Name or Material 2-Pentadecanone
EINECSNumber 219-064-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.