Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentadecanone 95.0+%, TCI America™
SDP

Catalog No. P106325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P106325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P106325G Supplier TCI America Supplier No. P106325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2345-28-0
Molecular Formula C15H30O
Molecular Weight (g/mol) 226.404
MDL Number MFCD00053712
InChI Key CJPNOLIZCWDHJK-UHFFFAOYSA-N
Synonym 2-pentadecanone, methyl tridecyl ketone, pentadecanone, unii-b2q48j997n, fema no. 3724, 2-pentandecanone, acmc-209g3f, 2-pentadecanone, fg, 2-pentadecanone, analytical standard
PubChem CID 61303
IUPAC Name pentadecan-2-one
SMILES CCCCCCCCCCCCCC(=O)C

Specifications

Melting Point 40°C
Color White
Boiling Point 162°C
Quantity 25 g
Formula Weight 226.40
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Pentadecanone
EINECSNumber (2)-0542
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.