Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Pentafluorophenoxy)ethanol 96.0+%, TCI America™
SDP

Catalog No. P12255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12255G Supplier TCI America Supplier No. P12255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2192-55-4
Molecular Formula C8H5F5O2
Molecular Weight (g/mol) 228.118
MDL Number MFCD00191478
InChI Key NPVVUYMVRISVGK-UHFFFAOYSA-N
PubChem CID 603435
IUPAC Name 2-(2,3,4,5,6-pentafluorophenoxy)ethanol
SMILES C(COC1=C(C(=C(C(=C1F)F)F)F)F)O

Specifications

Color Colorless
Boiling Point 81°C
Quantity 5 g
Formula Weight 228.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Pentafluorophenoxy)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.