Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Pentene (mixture of cis and trans)

Catalog No. 14377425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-377-425G 25 g
14-377-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-377-425G Supplier Sigma Aldrich Supplier No. 14377425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Pentene undergoes addition reaction with phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl)alkane adducts.2-Pentene (mixture of cis and trans) was used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

Specifications

CAS 109-68-2
Density 0.65 g/mL (at 25°C (literature))
Boiling Point 37°C (lit.)
Molecular Formula C5H10
Linear Formula CH3CH2CH=CHCH3
Refractive Index n20/D 1.38 (literature)
MDL Number MFCD00009384
Quantity 25 g
Molecular Weight (g/mol) 70.13
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.