Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Pentylfuran | 3777-69-3 | 25 G
SDP

Catalog No. 5000422037
Encompass_Preferred

2-Pentylfuran is a compound isolated from steam volatile oils obtained from potatoes at atmospheric pressure and is identified as a phytohormone. It has been observed to promote the growth of Arabidopsis thaliana at low concentrations while inhibiting plant growth at higher concentrations.

  • Phytohormone activity
  • Isolated from plant sources including potatoes
  • Promotes Arabidopsis thaliana growth at low concentrations
  • Inhibits Arabidopsis thaliana growth at high concentrations
  • Molecular formula: C9H14O
  • Molecular weight: 138.21
  • Appearance: Colorless to light yellow liquid
  • Recommended storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Recommended storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-22037 Supplier Medchemexpress LLC Supplier No. 25GHYN7398
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Pentylfuran is a compound isolated from steam volatile oils obtained from potatoes at atmospheric pressure and is identified as a phytohormone. It has been observed to promote the growth of Arabidopsis thaliana at low concentrations while inhibiting plant growth at higher concentrations.

  • Phytohormone activity
  • Isolated from plant sources including potatoes
  • Promotes Arabidopsis thaliana growth at low concentrations
  • Inhibits Arabidopsis thaliana growth at high concentrations
  • Molecular formula: C9H14O
  • Molecular weight: 138.21
  • Appearance: Colorless to light yellow liquid
  • Recommended storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Recommended storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.