Learn More
Medchemexpress LLC 2-Pentylfuran | 3777-69-3 | 5 G
SDP
Catalog No. 5000417652
Encompass_Preferred
2-Pentylfuran is a compound isolated from steam volatile oils obtained from potatoes at atmospheric pressure. It is also classified as a phytohormone, influencing plant growth.
  • Isolated from steam volatile oils obtained from potatoes
  • Functions as a phytohormone
  • Promotes Arabidopsis thaliana growth at low concentrations
  • Inhibits plant growth at high concentrations
  • Intended for research use only
Catalog No. 50-004-17652 Supplier Medchemexpress LLC Supplier No. HYN73985G
2-Pentylfuran is a compound isolated from steam volatile oils obtained from potatoes at atmospheric pressure. It is also classified as a phytohormone, influencing plant growth.
  • Isolated from steam volatile oils obtained from potatoes
  • Functions as a phytohormone
  • Promotes Arabidopsis thaliana growth at low concentrations
  • Inhibits plant growth at high concentrations
  • Intended for research use only