Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Pentylfuran 98 FG1KG
Catalog No. 502908391
Encompass
2-Pentylfuran is a volatile heterocyclic compound mainly associated with the flavor of reverted soybean oil. It is reported to be formed by the auto-oxidation of linolenic acid.
Catalog No. 50-290-8391 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3317081KGK
2-Pentylfuran is a volatile heterocyclic compound mainly associated with the flavor of reverted soybean oil. It is reported to be formed by the auto-oxidation of linolenic acid.