Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyaniline, 98%, Thermo Scientific Chemicals

Catalog No. p-3211736
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC373160250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC373160250 Supplier Thermo Scientific Chemicals Supplier No. 373160250
Only null left
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2688-84-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.22
MDL Number MFCD00035765
InChI Key NMFFUUFPJJOWHK-UHFFFAOYSA-N
Synonym 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
PubChem CID 75899
IUPAC Name 2-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color Brown to Gray
Boiling Point 170°C (18.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OC6H4NH2
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 185.22
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Phenoxyaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 220-254-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.