Learn More
2-Phenoxyethanol, 94%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.
Solubility
Miscible with water, ethanol, propanol and propylene glycol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 11°C to 14°C |
| Density | 1.102 |
| Boiling Point | 245°C |
| Flash Point | 121°C (249°F) |
| Assay Percent Range | 94% |
| Refractive Index | 1.537 |
| Linear Formula | C6H5O(CH2)2OH |
| Quantity | 5000 g |
| Beilstein | 1364011 |
| Merck Index | 14,7257 |
| Show More |
Safety and Handling
| Hazard Category | H302-H319 |
| Hazard Statement | GHS H Statement H302-H319 Harmful if swallowed. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P330-P501c |
| EINECSNumber | 204-589-7 |
| RTECSNumber | KM0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.