Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyethanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012814 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2814 5 mL
11-101-2813 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2814 Supplier MilliporeSigma Supelco Supplier No. 567535MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenoxyethanol is a compound, which belongs to the class of volatile organicshaving a faint rose like aromatic odor. It may find applications as a solvent in ball-point inks, resins and in organic synthesis. It may also be used in cosmetics as an antimicrobial agent and preservative.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 122-99-6
Density 1.107 g/mL (at 20°C)
Flash Point 121°C
Physical Form Neat
Linear Formula C6H5 OCH2 CH2 OH
MDL Number MFCD00002857
Melting Point 11°C to 13°C
Boiling Point 244°C to 246°C
Synonym Ethylene glycol monophenyl ether; PHE; PHE-G; PHE-S; PhG; Phenylglycol
Type Analytical Standard
Quantity 5 mL
Formula Weight 138.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.