Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyethanol, MP Biomedicals™

Catalog No. ICN15014880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
0.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15014880 100 mL
ICN15014890 0.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15014880 Supplier MP Biomedicals, Inc Supplier No. 0215014880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
PubChem CID 31236
ChEBI CHEBI:64275
IUPAC Name 2-phenoxyethanol
SMILES C1=CC=C(C=C1)OCCO

Specifications

Quantity 100 mL
Formula Weight 138.2
Chemical Name or Material 2-Phenoxyethanol
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.