Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyethanol Pract., 90%, Thermo Scientific™

Catalog No. AC417290010
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC417290010 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC417290010 Supplier Thermo Scientific Chemicals Supplier No. 417290010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 122-99-6
CAS Min % 88.0
CAS Max % 100.0
Density 1.1g/mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Molecular Formula C8H10O2
Refractive Index 1.5360 to 1.5400 (20°C, 589nm)
Linear Formula C6H5OCH2CH2OH
Quantity 1kg
Beilstein 06, 146
Merck Index 15, 7369
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OCCO
IUPAC Name 2-phenoxyethanol
Molecular Weight (g/mol) 138.166
PubChem CID 31236
ChEBI CHEBI:64275
Formula Weight 138.17
Percent Purity ≥88%
Chemical Name or Material 2-Phenoxyethanol, pract., 90%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-589-7
RTECSNumber KM0350000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.