Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyethyl Acetate 99.0+%, TCI America™
SDP

Catalog No. A093525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A093525G 25 g
A0935500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A093525G Supplier TCI America Supplier No. A093525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6192-44-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00059336
InChI Key WHFKYDMBUMLWDA-UHFFFAOYSA-N
Synonym Acetic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acetate
PubChem CID 22568
IUPAC Name 2-phenoxyethyl acetate
SMILES CC(=O)OCCOC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 231°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethyl Acetate
EINECSNumber (3)-0564
RTECSNumber KM0525000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.