Learn More
Cayman Chemical 2-Phenethylamine (hydrochloride), Purity: ≥98%, CAS: 156-28-5, 50 mg
SDP
Catalog No. 502954522
Encompass_Preferred
A natural monoamine alkaloid that acts as neuromodulator in the central nervous system; stimulates the release of norepinephrine and dopamine and is metabolized by monoamine oxidases as well as other enzymes; intended for forensic and research applications
Catalog No. 50-295-4522 Supplier Cayman Chemical Supplier No. 1420650 MG
A natural monoamine alkaloid that acts as neuromodulator in the central nervous system; stimulates the release of norepinephrine and dopamine and is metabolized by monoamine oxidases as well as other enzymes; intended for forensic and research applications