Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-1,3-thiazole-4-carbonyl chloride, Thermo Scientific™

Catalog No. CC18302DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18302DE 5 g
CC18302CB 250 mg
CC18302DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC18302DE Supplier Thermo Scientific Chemicals Supplier No. CC18302DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36094-04-9
Molecular Formula C10H6ClNOS
Molecular Weight (g/mol) 223.674
InChI Key ZZFOIDMEHXGBKS-UHFFFAOYSA-N
Synonym 2-phenylthiazole-4-carbonyl chloride, 2-phenyl-thiazole-4-carbonyl chloride, 4-thiazolecarbonylchloride, 2-phenyl
PubChem CID 2776472
IUPAC Name 2-phenyl-1,3-thiazole-4-carbonyl chloride
SMILES C1=CC=C(C=C1)C2=NC(=CS2)C(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 223.68
Percent Purity 95%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.