Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-2-(2-piperidyl)acetamide Hydrate 97.0+%, TCI America™
SDP

Catalog No. P17975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17975G 5 g
P179725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17975G Supplier TCI America Supplier No. P17975G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19395-39-2
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
MDL Number MFCD06656234
InChI Key LJLMNWPXAYKPGV-UHFFFAOYSA-N
PubChem CID 86862
IUPAC Name 2-phenyl-2-piperidin-2-ylacetamide
SMILES C1CCNC(C1)C(C2=CC=CC=C2)C(=O)N

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.30
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-2-(2-piperidyl)acetamide Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.