Learn More
Medchemexpress LLC 2-Phenyl-2-(2-pyridyl)acetonitrile | 5005-36-7 | 100.0% | 194.23 | 100 MG
SDP
Catalog No. 5000366420
Encompass_Preferred
2-Phenyl-2-(2-pyridyl)acetonitrile is identified as the major metabolite of SC 15396, an antigastrin that inhibits gastric secretion. It is metabolized by the supernatant fraction of rat liver homogenate.
  • Major metabolite of SC 15396
  • Metabolized by rat liver homogenate supernatant fraction
  • Antigastrin
  • Inhibits gastric secretion
  • Off-white to light brown solid
  • High purity
Catalog No. 50-003-66420 Supplier Medchemexpress LLC Supplier No. HYW009934100MG
2-Phenyl-2-(2-pyridyl)acetonitrile is identified as the major metabolite of SC 15396, an antigastrin that inhibits gastric secretion. It is metabolized by the supernatant fraction of rat liver homogenate.
  • Major metabolite of SC 15396
  • Metabolized by rat liver homogenate supernatant fraction
  • Antigastrin
  • Inhibits gastric secretion
  • Off-white to light brown solid
  • High purity