Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Phenyl-2-(2-pyridyl)acetonitrile | 5005-36-7 | 100.0% | 194.23 | 500 MG
SDP

Catalog No. 5000366885
Encompass_Preferred

2-Phenyl-2-(2-pyridyl)acetonitrile is the major metabolite of SC 15396, metabolized by the supernatant fraction of rat liver homogenate. SC 15396 is an antigastrin that inhibits gastric secretion.

  • Appearance: Off-white to light brown solid
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • Solvent storage at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)
  • Documentation such as data sheet, COA, SDS, handling instructions, HNMR, and LCMS are available

Catalog No. 50-003-66885 Supplier Medchemexpress LLC Supplier No. HYW009934500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenyl-2-(2-pyridyl)acetonitrile is the major metabolite of SC 15396, metabolized by the supernatant fraction of rat liver homogenate. SC 15396 is an antigastrin that inhibits gastric secretion.

  • Appearance: Off-white to light brown solid
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • Solvent storage at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)
  • Documentation such as data sheet, COA, SDS, handling instructions, HNMR, and LCMS are available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.