Learn More
Medchemexpress LLC 2-phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 | C9H12O | 25 G
SDP
Catalog No. 5000303567
Encompass_Preferred
2-Phenyl-2-propanol is an aromatic tertiary alcohol used as an intermediate in organic synthesis. It is commonly used in the preparation of diisopropylbenzene peroxide, and as a building block for fragrance and cosmetic additive synthesis as well as general laboratory reagents.
Catalog No. 50-003-03567 Supplier Medchemexpress LLC Supplier No. 25GHYW015810
2-Phenyl-2-propanol is an aromatic tertiary alcohol used as an intermediate in organic synthesis. It is commonly used in the preparation of diisopropylbenzene peroxide, and as a building block for fragrance and cosmetic additive synthesis as well as general laboratory reagents.