Learn More
Medchemexpress LLC 2-phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 g/mol | C9H12O | 1 ML
SDP
Catalog No. 5000300698
Encompass_Preferred
2-Phenyl-2-propanol is an aromatic secondary alcohol (CAS 617-94-7) used as a synthetic intermediate and reagent in organic chemistry. It is applied in the preparation of peroxides, and as a precursor for fragrance and cosmetic additives; supplied in both solution and bulk powder formats to suit research and production needs.
  • Used as a synthetic intermediate for peroxide and fragrance precursors.
  • Suitable for organic synthesis applications and reagent-grade reactions.
  • Available as solution (for example, 10 mM, 1 mL) and in multiple powder sizes for scale flexibility.
  • Soluble in common solvents such as DMSO (reported solubility ~100 mg/mL).
  • High-purity grades available to support sensitive syntheses.
Catalog No. 50-003-00698 Supplier Medchemexpress LLC Supplier No. HYW01581010MM/1ML
2-Phenyl-2-propanol is an aromatic secondary alcohol (CAS 617-94-7) used as a synthetic intermediate and reagent in organic chemistry. It is applied in the preparation of peroxides, and as a precursor for fragrance and cosmetic additives; supplied in both solution and bulk powder formats to suit research and production needs.
  • Used as a synthetic intermediate for peroxide and fragrance precursors.
  • Suitable for organic synthesis applications and reagent-grade reactions.
  • Available as solution (for example, 10 mM, 1 mL) and in multiple powder sizes for scale flexibility.
  • Soluble in common solvents such as DMSO (reported solubility ~100 mg/mL).
  • High-purity grades available to support sensitive syntheses.