Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 g/mol | C9H12O | 250 G
SDP

Catalog No. 5000302834
Encompass_Preferred

2-Phenyl-2-propanol is an aromatic secondary alcohol used as a chemical intermediate and as an additive in fragrances and cosmetics. It is supplied as a high-purity research reagent for organic synthesis and R&D applications, with physical properties that make it solid at lower temperatures and liquid above its melting point.

  • High purity: 99.54%.
  • Molecular formula: C9H12O; molecular weight: 136.19.
  • Physical form: colorless to off-white; solid below 36 °C, liquid above 36 °C.
  • Available pack sizes range from small (5 g) to larger quantities (250 g, 500 g) for scale flexibility.
  • Suitable as an intermediate in organic synthesis and as a fragrance/cosmetic additive.
  • Product datasheet provided for specifications and handling information.

Catalog No. 50-003-02834 Supplier Medchemexpress LLC Supplier No. 250GHYW015810
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenyl-2-propanol is an aromatic secondary alcohol used as a chemical intermediate and as an additive in fragrances and cosmetics. It is supplied as a high-purity research reagent for organic synthesis and R&D applications, with physical properties that make it solid at lower temperatures and liquid above its melting point.

  • High purity: 99.54%.
  • Molecular formula: C9H12O; molecular weight: 136.19.
  • Physical form: colorless to off-white; solid below 36 °C, liquid above 36 °C.
  • Available pack sizes range from small (5 g) to larger quantities (250 g, 500 g) for scale flexibility.
  • Suitable as an intermediate in organic synthesis and as a fragrance/cosmetic additive.
  • Product datasheet provided for specifications and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.