Learn More
Medchemexpress LLC 2-Phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 g/mol | C9H12O | 5 G
SDP
Catalog No. 5000300024
Encompass_Preferred
2-Phenyl-2-propanol is an aromatic secondary alcohol (C9H12O, MW 136.19 g/mol) commonly used as a synthetic intermediate in organic chemistry and as an additive in fragrances and cosmetic formulations. It is provided at high purity, available in laboratory pack sizes, and requires controlled storage to preserve stability and shelf life.
  • High purity (reported 99.54%) suitable for synthetic applications.
  • Useful intermediate for peroxide formation and other organic transformations.
  • Applicable as an additive in fragrance and cosmetic formulations.
  • Available in multiple pack sizes to support lab-scale and preparative work.
  • Solid below ~36°C and liquid above ~36°C; handle under appropriate temperature conditions.
  • Recommended refrigerated storage prolongs shelf life.
Catalog No. 50-003-00024 Supplier Medchemexpress LLC Supplier No. HYW0158105G
2-Phenyl-2-propanol is an aromatic secondary alcohol (C9H12O, MW 136.19 g/mol) commonly used as a synthetic intermediate in organic chemistry and as an additive in fragrances and cosmetic formulations. It is provided at high purity, available in laboratory pack sizes, and requires controlled storage to preserve stability and shelf life.
  • High purity (reported 99.54%) suitable for synthetic applications.
  • Useful intermediate for peroxide formation and other organic transformations.
  • Applicable as an additive in fragrance and cosmetic formulations.
  • Available in multiple pack sizes to support lab-scale and preparative work.
  • Solid below ~36°C and liquid above ~36°C; handle under appropriate temperature conditions.
  • Recommended refrigerated storage prolongs shelf life.