Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Phenyl-2-propanol | 617-94-7 | MFCD00004456 | 99.5% | 136.19 g/mol | C9H12O | 50 G
SDP

Catalog No. 5000300303
Encompass_Preferred

2-Phenyl-2-propanol is an aromatic tertiary alcohol used as an intermediate in organic synthesis and as an additive in fragrances and cosmetics. It is supplied as a high-purity reagent intended for laboratory research and synthetic applications, with recommended handling and storage to preserve stability.

  • Purity 99.54%.
  • CAS number 617-94-7.
  • Used as an intermediate in organic synthesis and as an additive in fragrances and cosmetics.
  • Recommended solvent DMSO (100 mg/mL).
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available documentation includes datasheet, certificate of analysis, and SDS.
  • Pack size 50 g.

Catalog No. 50-003-00303 Supplier Medchemexpress LLC Supplier No. HYW01581050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenyl-2-propanol is an aromatic tertiary alcohol used as an intermediate in organic synthesis and as an additive in fragrances and cosmetics. It is supplied as a high-purity reagent intended for laboratory research and synthetic applications, with recommended handling and storage to preserve stability.

  • Purity 99.54%.
  • CAS number 617-94-7.
  • Used as an intermediate in organic synthesis and as an additive in fragrances and cosmetics.
  • Recommended solvent DMSO (100 mg/mL).
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available documentation includes datasheet, certificate of analysis, and SDS.
  • Pack size 50 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.