Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-4-penten-2-ol 97.0+%, TCI America™
SDP

Catalog No. P13305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13305G Supplier TCI America Supplier No. P13305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4743-74-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00060886
InChI Key BELRNEPYFJNSPN-UHFFFAOYSA-N
Synonym 2-phenyl-4-penten-2-ol, 2-phenyl 4-penten-2-ol, allyl methyl phenyl carbinol, phenyl-methyl-allyl-carbinol
PubChem CID 143761
IUPAC Name 2-phenylpent-4-en-2-ol
SMILES CC(CC=C)(C1=CC=CC=C1)O

Specifications

Boiling Point 100°C
Quantity 5 g
Formula Weight 162.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenyl-4-penten-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.