Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-4-quinolinecarboxylic acid, 99%, Thermo Scientific™

Catalog No. AC188142500
Change view
Click to view available options
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC188142500 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188142500 Supplier Thermo Scientific Chemicals Supplier No. 188142500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 132-60-5
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C16H11NO2
Quantity 250g
MDL Number MFCD00006750
Beilstein 22, 103
Merck Index 15, 2291
Synonym cinchophen, tervalon, 2-phenyl-4-quinolinecarboxylic acid, cinchophene, cinconal, phenoquin, quinofen, quinophan, quinophen, atofan
InChI Key YTRMTPPVNRALON-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)C(=O)O
IUPAC Name 2-phenylquinoline-4-carboxylic acid
Molecular Weight (g/mol) 249.269
PubChem CID 8593
Formula Weight 249.27
Percent Purity ≥98.5%
Chemical Name or Material 2-Phenyl-4-quinolinecarboxylic acid, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-067-1
RTECSNumber GD4025000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.