Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-L-glycinamide hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH6575103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6575103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6575103 Supplier Thermo Scientific Chemicals Supplier No. H6575103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60079-51-8
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.639
MDL Number MFCD00270568
InChI Key LXOAFHGMXCFTOU-FJXQXJEOSA-N
Synonym s-2-amino-2-phenylacetamide hydrochloride, l-phenylglycine amide hydrochloride, h-phg-nh2 hcl, h-phg-nh2.hcl, 2s-2-amino-2-phenylacetamide hydrochloride, hpphg-nh2 hcl, h-phg-nh2 . hcl, phenylglycine amide hydrochloride, s-2-amino-2-phenyl-acetamide hydrochloride, alphas-alpha-amino-benzeneacetamide hydrochloride
PubChem CID 22850726
IUPAC Name (2S)-2-amino-2-phenylacetamide;hydrochloride
SMILES C1=CC=C(C=C1)C(C(=O)N)N.Cl

Specifications

Quantity 1 g
Beilstein 4823667
Formula Weight 186.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Phenyl-L-glycinamide hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.