Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 g·mol⁻¹ | C8H9NO | 1 ML
SDP

Catalog No. 5000299901
Encompass_Preferred

2-Phenylacetamide is a high-purity small-molecule p38 MAPK inhibitor intended for biochemical and cellular research. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO, with characterized identity and storage guidance for laboratory use.

  • High purity, 99.92%.
  • Available as solid or 10 mM solution in DMSO for ready-to-use assays.
  • Molecular formula C8H9NO and molecular weight 135.17 g·mol⁻¹.
  • Characterized by SMILES and standard identifiers, including CAS 103-81-1.
  • Storage recommendations provided for powder and solution formats.

Catalog No. 50-002-99901 Supplier Medchemexpress LLC Supplier No. HYW01819710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylacetamide is a high-purity small-molecule p38 MAPK inhibitor intended for biochemical and cellular research. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO, with characterized identity and storage guidance for laboratory use.

  • High purity, 99.92%.
  • Available as solid or 10 mM solution in DMSO for ready-to-use assays.
  • Molecular formula C8H9NO and molecular weight 135.17 g·mol⁻¹.
  • Characterized by SMILES and standard identifiers, including CAS 103-81-1.
  • Storage recommendations provided for powder and solution formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.