Learn More
Medchemexpress LLC 2-phenylacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 g/mol | C8H9NO | 10 G
SDP
Catalog No. 5000301436
Encompass_Preferred
2-Phenylacetamide (CAS 103-81-1) is a small-molecule research compound (molecular formula C8H9NO, molecular weight 135.17 g/mol) reported to inhibit the p38 MAPK signaling pathway and to exhibit anti-inflammatory, antioxidant, anti-hypertensive, and anti-fibrosis effects. It is supplied as a solid at research-grade purity for biochemical and pharmacological studies.
  • High purity suitable for research applications.
  • Reported p38 MAPK pathway inhibitory activity.
  • Available in multiple pack sizes, including 10 g.
  • Supplied as a solid with molecular formula C8H9NO.
  • Useful for biochemical, pharmacological, and pathway-modulation studies.
Catalog No. 50-003-01436 Supplier Medchemexpress LLC Supplier No. HYW01819710G
2-Phenylacetamide (CAS 103-81-1) is a small-molecule research compound (molecular formula C8H9NO, molecular weight 135.17 g/mol) reported to inhibit the p38 MAPK signaling pathway and to exhibit anti-inflammatory, antioxidant, anti-hypertensive, and anti-fibrosis effects. It is supplied as a solid at research-grade purity for biochemical and pharmacological studies.
  • High purity suitable for research applications.
  • Reported p38 MAPK pathway inhibitory activity.
  • Available in multiple pack sizes, including 10 g.
  • Supplied as a solid with molecular formula C8H9NO.
  • Useful for biochemical, pharmacological, and pathway-modulation studies.