Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylacetamide | 103-81-1 | MFCD00059193 | 99.9% | 135.17 g/mol | C8H9NO | 25 G
SDP

Catalog No. 5000304165
Encompass_Preferred

2-Phenylacetamide (CAS 103-81-1) is a white to off-white solid used as an analytical standard, metabolite reference, and research reagent in studies of metabolic pathways, reactive oxygen species, and p38 MAPK-related assays. It offers high purity and well-documented analytical data for laboratory use.

  • High purity (99.92%) suitable for analytical and research applications.
  • White to off-white solid physical form.
  • Molecular formula C8H9NO; molecular weight 135.17 g/mol.
  • Soluble in DMSO (≈100 mg/mL); protocols provided for in vivo formulations.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years).
  • Comes with COA, SDS, and analytical data (HNMR, LCMS, RP-HPLC).

Catalog No. 50-003-04165 Supplier Medchemexpress LLC Supplier No. 25GHYW018197
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylacetamide (CAS 103-81-1) is a white to off-white solid used as an analytical standard, metabolite reference, and research reagent in studies of metabolic pathways, reactive oxygen species, and p38 MAPK-related assays. It offers high purity and well-documented analytical data for laboratory use.

  • High purity (99.92%) suitable for analytical and research applications.
  • White to off-white solid physical form.
  • Molecular formula C8H9NO; molecular weight 135.17 g/mol.
  • Soluble in DMSO (≈100 mg/mL); protocols provided for in vivo formulations.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years).
  • Comes with COA, SDS, and analytical data (HNMR, LCMS, RP-HPLC).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.