Learn More
Cayman Chemical 2-Phenylacetamide, Purity: ≥98%, CAS: 103-81-1, 10 g
SDP
Catalog No. 502911676
Encompass_Preferred
A metabolite of L-phenylalanine; increases the proliferation of MCF-7 cells; increases luteinizing hormone and estrogen levels and ERα and ERβ expression in the uterus of immature female mice; urinary levels are increased in male obese patients compared with non-obese males
Catalog No. 50-291-1676 Supplier Cayman Chemical Supplier No. 3636410 G
A metabolite of L-phenylalanine; increases the proliferation of MCF-7 cells; increases luteinizing hormone and estrogen levels and ERα and ERβ expression in the uterus of immature female mice; urinary levels are increased in male obese patients compared with non-obese males