Learn More
Medchemexpress LLC 2-phenylacetophenone | 451-40-1 | MFCD00003081 | 1mL
SDP
Catalog No. 5000265945
Encompass_Preferred
2-Phenylacetophenone has broad-spectrum efflux pump inhibition activity 2-Phenylacetophenone is a benzoin derivative used as a photoinitiator in vinyl polymerization[1]
Catalog No. 50-002-65945 Supplier Medchemexpress LLC Supplier No. HYY177710MM/1ML
2-Phenylacetophenone has broad-spectrum efflux pump inhibition activity 2-Phenylacetophenone is a benzoin derivative used as a photoinitiator in vinyl polymerization[1]