Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylanthraquinone 98.0+%, TCI America™
SDP

Catalog No. P25881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25881G 1 g
P25885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25881G Supplier TCI America Supplier No. P25881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6485-97-8
Molecular Formula C20H12O2
Molecular Weight (g/mol) 284.314
MDL Number MFCD00559262
InChI Key NTZCFGZBDDCNHI-UHFFFAOYSA-N
Synonym 2-phenylanthraquinone, 2-phenylanthra-9,10-quinone, 2-phenyl-9,10-anthracenedione, 2-phenylanthrachinon, acmc-20annp, 9, 2-phenyl, 2-phenyl-9,10-anthraquinone, 9,10-anthracenedione,2-phenyl
PubChem CID 313995
IUPAC Name 2-phenylanthracene-9,10-dione
SMILES C1=CC=C(C=C1)C2=CC3=C(C=C2)C(=O)C4=CC=CC=C4C3=O

Specifications

Melting Point 162°C
Color Yellow
Quantity 1 g
Formula Weight 284.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.