Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Phenylbutyric acid

Catalog No. P20955100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20955100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20955100G Supplier Sigma Aldrich Supplier No. P20955100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-methoxyfuran forms cycloadducts with maleic anhydride and n-methylmaleimide
  • It undergoes friedel-crafts reaction with nitroalkenes catalyzed by diphenylamine-tethered bis(oxazoline)-zn(otf)2 complexes.2-methoxyfuran was used to study the hydrolysis of furan derivative by acid catalysis

Specifications

CAS 90-27-7
Melting Point 39°C to 42°C (lit.)
Boiling Point 270°C to 272°C (lit.)
Molecular Formula C10H12O2
Linear Formula CH3CH2CH(C6H5)COOH
MDL Number MFCD00002667
Quantity 5 g
Synonym (+/-)-2-Phenylbutyric acid; alpha-Ethylphenylacetic acid
Molecular Weight (g/mol) 164.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.